Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C8orf33 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318500JF549
Description
C8orf33 Polyclonal antibody specifically detects C8orf33 in Human samples. It is validated for Western BlotSpecifications
C8orf33 | |
Polyclonal | |
Western Blot | |
chromosome 8 open reading frame 33, FLJ20989, hypothetical protein LOC65265 | |
Recognizes region between residues Between between 25 and 75.. | |
0.1 mL | |
Stem Cell Markers | |
65265 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction