Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C8orf34 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565760 Supplier Invitrogen™ Supplier No. PA565760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LGKALENLSRS IAISDELDKE TVTFNSSLLR PRVIGEWIGR EENDADPLAA EMLQPPIPRS KNDQWESEDS GSSPAGSLKM EPKNKGLKQ Highest antigen sequence identity to the following orthologs - mouse 92%, rat 91%.

This gene encodes a protein that is related to the cyclic AMP dependent protein kinase regulators. Naturally occurring mutations in this gene are associated with an increased risk for severe toxicities, such as diarrhea and neutropenia, in patients undergoing chemotherapeutic treatment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C8orf34
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C8orf34
Gene Accession No. Q49A92
Gene Alias C8orf34; chromosome 8 open reading frame 34; protein VEST-1; Uncharacterized protein C8orf34; VEST1; VEST-1; vestibule 1; vestibule-1 protein
Gene Symbols C8orf34
Host Species Rabbit
Immunogen Recombinant Human C8orf34. Recombinant protein control fragment (Product #RP-106440).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.