Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C8orf4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983637
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3637 Supplier Bioss Supplier No. BS15289R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a small, monomeric, predominantly unstructured protein that functions as a positive regulator of the Wnt/beta-catenin signaling pathway. This protein interacts with a repressor of beta-catenin mediated transcription at nuclear speckles. It is thought to competitively block interactions of the repressor with beta-catenin, resulting in up-regulation of beta-catenin target genes. The encoded protein may also play a role in the NF-kappaB and ERK1/2 signaling pathways. Expression of this gene may play a role in the proliferation of several types of cancer including thyroid cancer, breast cancer and hematological malignancies.

Specifications

Antigen C8orf4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Tcim
Gene Accession No. Q9NR00
Gene Alias 1110065B09Rik; 1810011O10Rik; AW121743; AW321058; BOS_23908; C16H8orf4; C27H8orf4; C8orf4; chromosome 16 open reading frame, human C8orf4; chromosome 27 open reading frame, human C8orf4; chromosome 8 open reading frame 4; human thyroid cancer 1; LOC100349447; protein C8orf4; RIKEN cDNA 1810011O10 gene; Tc1; TC-1; TCIM; Thyroid cancer protein 1; thyroid cancer-1; thyroid cancer-1-like; Transcriptional and immune response regulator; uncharacterized protein C8orf4 homolog
Gene Symbols Tcim
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C8orf4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56892
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.