Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C8orf45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569931
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569931 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569931 Supplier Thermo Scientific Supplier No. PA569931
Add to Cart
Add to Cart

C8orf45 Polyclonal Antibody

C8orf45 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen C8orf45
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene MCMDC2
Gene Accession No. Q4G0Z9
Gene Alias 6030422M02Rik; C29H8orf45; C8orf45; LOW QUALITY PROTEIN: minichromosome maintenance domain-containing protein 2; MCM domain-containing protein 2; MCM domain-containing protein C8orf45; MCM domain-containing protein C8orf45 homolog; MCMDC2; minichromosome maintenance domain containing 2; minichromosome maintenance domain-containing protein 2
Gene Symbols MCMDC2
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human C8orf45.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157777
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.