Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C8orf46 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567375
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567375 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567375 Supplier Invitrogen™ Supplier No. PA567375
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QGAEASLPLTG SASCGVPGIL RKMWTRHKKK SEYVGATNSA FEA Highest antigen sequence identity to the following orthologs - mouse 98%, rat 100%.

Required for neurogenesis in the neural plate and retina. Strongly cooperates with neural bHLH factors to promote neurogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C8orf46
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C8orf46
Gene Accession No. Q8TAG6
Gene Alias C8orf46; chromosome 8 open reading frame 46; uncharacterized protein C8orf46; Vexin; VXN
Gene Symbols C8orf46
Host Species Rabbit
Immunogen Recombinant Human C8orf46. Recombinant protein control fragment (Product #RP-108036).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254778
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.