Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C8orf55 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA582823
Description
Immunogen sequence: LLGWDDRAFY LEARFVSLRD GFVCALLRFR QHLLGTSPER VVQHLCQRRV EPPELPADLQ HWISYNEASS QLLRMESGLS DVTKDQ.
DSCD75 is a 208 amino acid membrane protein with one transmembrane helix. Current testing suggests that the cDNA of DSCD75 may be expressed by osteogenic human mesenchymal stem cells. DSCD75 may be involved in some transporter activities.
Specifications
C8orf55 | |
Polyclonal | |
Unconjugated | |
THEM6 | |
4930572J05Rik; C13H8orf55; C14H8orf55; C8orf55; DSCD75; mesenchymal stem cell protein DSCD75; mesenchymal stem cell protein DSCD75 homolog; protein THEM6; PSEC0098; RGD1359378; Them6; Thioesterase superfamily member 6; UPF0670 protein C8orf55; UPF0670 protein C8orf55 homolog; UPF0670 protein THEM6; UPF0670 protein THEM6 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51337 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WUY1 | |
THEM6 | |
Recombinant protein corresponding to Human THEM6. Recombinant protein control fragment (Product #RP-93784). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction