Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C8orf55 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983645
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3645 Supplier Bioss Supplier No. BS15296R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DSCD75 is a 208 amino acid membrane protein with one transmembrane helix. Current testing suggests that the cDNA of DSCD75 may be expressed by osteogenic human mesenchymal stem cells. DSCD75 may be involved in some transporter activities.

Specifications

Antigen C8orf55
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene THEM6
Gene Accession No. Q8WUY1
Gene Alias 4930572J05Rik; BOS_13668; C13H8orf55; C14H8orf55; C8orf55; DSCD75; Mesenchymal stem cell protein DSCD75; mesenchymal stem cell protein DSCD75 homolog; Protein THEM6; PSEC0098; RGD1359378; THEM6; Thioesterase superfamily member 6; UPF0670 protein C8orf55; UPF0670 protein C8orf55 homolog; UPF0670 protein THEM6; UPF0670 protein THEM6 homolog
Gene Symbols THEM6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C8orf55.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51337
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.