Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C8orf58 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555813
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555813 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555813 Supplier Invitrogen™ Supplier No. PA555813
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEQMARLQQL YLQLRIQRPP GDPGEEESTR APLPSPLHTP GNRGQGPWEL LSQTEHTGAK AASPPKVEVP SANPPRLPET Highest antigen sequence identity to the following orthologs: Mouse - 57%, Rat - 60%.

C8orf58 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C8orf58
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C8orf58
Gene Accession No. Q8NAV2
Gene Alias C8orf58; chromosome 8 open reading frame 58; uncharacterized protein C8orf58
Gene Symbols C8orf58
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C8orf58. Recombinant protein control fragment (Product #RP-93857).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 541565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.