Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9 Neoantigen Monoclonal Antibody (WU13-15)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549003
Description
C9 Neoantigen Monoclonal Antibody for Western Blot, ICC/IF, IHC (F), IHC (P), Flow, IA
C9 neoantigen is a target gene that is involved in the immune system's response to cancer cells. Neoantigens are proteins that are produced by cancer cells and are not present in normal cells. C9 neoantigen is a protein fragment that is produced when the C9 protein is broken down by enzymes in the cancer cells. The immune system recognizes these neoantigens as foreign and mounts an attack against the cancer cells. C9 neoantigen is being studied as a potential target for cancer immunotherapy, which involves using the patient's own immune system to fight cancer.
Specifications
C9 Neoantigen | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
C9 | |
The monoclonal antibody WU13-15 recognizes a neoepitope on the 61 kDa complement component C9, integrated in the terminal complement complex (TCC). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Inhibition Assays, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
WU13-15 | |
Unconjugated | |
P29466, P48729, Q8N752 | |
Mouse | |
Protein G | |
RUO | |
122011, 1452, 834 | |
4°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction