Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ARHGEF39 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549538
Description
Promotes cell proliferation.
Specifications
ARHGEF39 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q8N4T4 | |
ARHGEF39 | |
KLH conjugated synthetic peptide between 180-211 amino acids from the Central region of human C9orf100. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ARHGEF39 | |
Rho guanine nucleotide exchange factor 39, ARHGEF39, C9orf100 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84904 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction