Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf114 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582808
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582808 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582808 Supplier Invitrogen™ Supplier No. PA582808
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QYLRKAFFPK HQDLQFAGLL NPLDSPHHMR QDEESEFREG IVVDRPTRPG HGSFVNCGMK KEVKIDKNLE PGLRVTVRLN QQQHPDCKTY HGKVVSSQDP RTKAGLYWGY TVRLASC Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 91%.

C9orf114 is a protein coding gene. Required for association of the centrosomes with the poles of the bipolar mitotic spindle during metaphase. Also involved in chromosome alignment. May promote centrosome maturation probably by recruiting A-kinase anchor protein AKAP9 to centrosomes in early mitosis. Binds specifically to miRNA MIR145 hairpin, regulates MIR145 expression at a postranscriptional level.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf114
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPOUT1
Gene Accession No. Q5T280
Gene Alias 9930028P05; AL033358; C9orf114; CENP-32; Centromere protein 32; D2Wsu81e; DNA segment, Chr 2, Wayne State University 81, expressed; HSPC109; Kinetochore-associated protein; LOC499770; Putative methyltransferase C9orf114; putative methyltransferase C9orf114 homolog; similar to LOC495800 protein; SPOUT domain containing methyltransferase 1; SPOUT domain-containing methyltransferase 1; Spout1; uncharacterized protein C9orf114 homolog
Gene Symbols SPOUT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SPOUT1. Recombinant protein control fragment (Product #RP-93104).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51490
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.