Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf115 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983658
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3658 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3658 Supplier Bioss Supplier No. BS15309R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PTRH1 (Peptidyl-TRNA Hydrolase 1 Homolog) is a Protein Coding gene. Diseases associated with PTRH1 include Vascular Skin Disease and Early Infantile Epileptic Encephalopathy. Gene Ontology (GO) annotations related to this gene include aminoacyl-tRNA hydrolase activity.

Specifications

Antigen C9orf115
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PTRH1
Gene Accession No. Q86Y79
Gene Symbols PTRH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C9orf115.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138428
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.