Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf116 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570976 Supplier Invitrogen™ Supplier No. PA570976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Guinea Pig: 79%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 86%; Zebrafish: 90%.

C9orf116 (Chromosome 9 Open Reading Frame 116) is a Protein Coding gene. An important paralog of this gene isC15orf65.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf116
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C9orf116
Gene Accession No. Q5BN46
Gene Alias C9orf116; chromosome 9 open reading frame 116; p53-induced expression 1 in Rb-/- cells; p53-induced expression in RB-null cells protein 1; pierce 1; PIERCE1; RbEST47; UPF0691 protein C9orf116
Gene Symbols C9orf116
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C9orf116.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138162
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.