Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf116 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554408
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554408 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554408 Supplier Invitrogen™ Supplier No. PA554408
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PTVHEMPKVF YPNSNKFSQQ LAAGGMFRNN TLNVYLEKSI VTGPDNCITS CDRLNFHPSY NINRPSICD Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 71%.

C9orf116 (Chromosome 9 Open Reading Frame 116) is a Protein Coding gene. An important paralog of this gene isC15orf65.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf116
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C9orf116
Gene Accession No. Q5BN46
Gene Alias C9orf116; chromosome 9 open reading frame 116; p53-induced expression 1 in Rb-/- cells; p53-induced expression in RB-null cells protein 1; pierce 1; PIERCE1; RbEST47; UPF0691 protein C9orf116
Gene Symbols C9orf116
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf116. Recombinant protein control fragment (Product #RP-93023).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138162
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.