Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf117 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554508 Supplier Invitrogen™ Supplier No. PA554508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DNQALKSQRD QLSLQLEQQQ VDLQRLQQEL ANEQKVRASL EAALVQATSF LQNILQMHRD EEDSDVDVTF QPWHKEMLQQ LLVMLSST Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 55%.

Specifically required during spermatogenesis for flagellum morphogenesis and sperm motility. May be required to suppress the formation of supernumerary axonemes and ensure a correct ultrastructure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf117
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP157
Gene Accession No. Q5JU67
Gene Alias 1700019L03Rik; C25H9orf117; C9H9orf117; C9orf117; Cfap157; cilia and flagella associated protein 157; cilia- and flagella-associated protein 157; LOC499781; LOW QUALITY PROTEIN: uncharacterized protein C9orf117 homolog; RIKEN cDNA 1700019L03 gene; similar to CG17122-PA; uncharacterized protein C9orf117; uncharacterized protein C9orf117 homolog
Gene Symbols CFAP157
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf117. Recombinant protein control fragment (Product #RP-93024).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286207
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.