Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf117 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570012
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570012 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570012 Supplier Invitrogen™ Supplier No. PA570012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.

Specifically required during spermatogenesis for flagellum morphogenesis and sperm motility. May be required to suppress the formation of supernumerary axonemes and ensure a correct ultrastructure.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf117
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene CFAP157
Gene Accession No. Q5JU67
Gene Alias 1700019L03Rik; C25H9orf117; C9H9orf117; C9orf117; Cfap157; cilia and flagella associated protein 157; cilia- and flagella-associated protein 157; LOC499781; LOW QUALITY PROTEIN: uncharacterized protein C9orf117 homolog; RIKEN cDNA 1700019L03 gene; similar to CG17122-PA; uncharacterized protein C9orf117; uncharacterized protein C9orf117 homolog
Gene Symbols CFAP157
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human C9orf117.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286207
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.