Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf126 Monoclonal Antibody (7-A4)
GREENER_CHOICE

Catalog No. PIMA532873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532873 Supplier Invitrogen™ Supplier No. MA532873
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C9orf126 Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

This gene encodes a regulator of cell migration. The encoded protein appears to function in the RhoA (ras homolog gene family, member A)-Dia1 (diaphanous homolog 1) signal transduction pathway. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf126
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7-A4
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene SCAI
Gene Accession No. Q8N9R8
Gene Alias 9330112M16; A930041I02Rik; AI662729; C9orf126; NET40; protein SCAI; SCAI; suppressor of cancer cell invasion; suppressor of cancer cell invasion protein
Gene Symbols SCAI
Host Species Mouse
Immunogen Recombinant protein within Human SCAI aa 1-606.
Purification Method Protein L
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286205
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.