Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf126 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143759
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143759 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143759 Supplier Invitrogen™ Supplier No. PA5143759
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects a significant 66 kDa* protein corresponding to the molecular mass of SCAI on SDS-PAGE immunoblots of mouse brain and human A431 cells. In brain, the antibody also detects 53 and 41 kDa bands.

This gene encodes a regulator of cell migration. The encoded protein appears to function in the RhoA (ras homolog gene family, member A)-Dia1 (diaphanous homolog 1) signal transduction pathway. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf126
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene SCAI
Gene Accession No. Q8C8N2, Q8N9R8
Gene Alias 9330112M16; A930041I02Rik; AI662729; C9orf126; NET40; protein SCAI; SCAI; suppressor of cancer cell invasion; suppressor of cancer cell invasion protein
Gene Symbols SCAI
Host Species Rabbit
Immunogen SCAI synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of mouse SCAI. This peptide sequence is highly conserved in human, rat, mouse, and chicken SCAI.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286205, 320271, 690538
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.