Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf140 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983663
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3663 Supplier Bioss Supplier No. BS15314R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen C9orf140
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SAPCD2
Gene Accession No. Q86UD0
Gene Alias 2010317E24Rik; C9orf140; p42.3; protein C9orf140; SAPCD2; suppressor APC domain containing 2; Suppressor APC domain-containing protein 2; TS/MDEP; Tumor specificity and mitosis phase-dependent expression protein
Gene Symbols SAPCD2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C9orf140.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89958
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.