Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf142 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560967
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560967 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560967 Supplier Invitrogen™ Supplier No. PA560967
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSKVPGPEAA PRLRALTLGL AKRVWSLERR LAAAEETAVS PRKSPRPAGP QLFLPDPDPQ RGGPGPGVRR RCPGESLINP GFKSKKPAGG VDFDET Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

The protein encoded by this gene plays a role in the nonhomologous end joining (NHEJ) pathway of DNA double-strand break repair. The encoded protein may function to stabilize the Ku70/Ku80 heterodimer to facilitate the assembly and maintain the stability of the NHEJ complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf142
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.30 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAXX
Gene Accession No. Q9BUH6
Gene Alias BC029214; C9orf142; cDNA sequence BC029214; chromosome 9 open reading frame 142; D930050G13Rik; Paralog of XRCC4 and XLF; Paxx; Protein PAXX; RGD1306215; similar to hypothetical protein MGC36831; uncharacterized protein C9orf142 homolog; XLS; XRCC4-like small protein
Gene Symbols PAXX
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf142. Recombinant protein control fragment (Product #RP-98479).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286257
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.