Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf142 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114924 Supplier Invitrogen™ Supplier No. PA5114924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C9orf142.

The protein encoded by this gene plays a role in the nonhomologous end joining (NHEJ) pathway of DNA double-strand break repair. The encoded protein may function to stabilize the Ku70/Ku80 heterodimer to facilitate the assembly and maintain the stability of the NHEJ complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf142
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene PAXX
Gene Accession No. Q8K0Y7, Q9BUH6
Gene Alias BC029214; C9orf142; cDNA sequence BC029214; chromosome 9 open reading frame 142; D930050G13Rik; Paralog of XRCC4 and XLF; Paxx; Protein PAXX; RGD1306215; similar to hypothetical protein MGC36831; uncharacterized protein C9orf142 homolog; XLS; XRCC4-like small protein
Gene Symbols PAXX
Host Species Rabbit
Immunogen A synthesized peptide derived from human C9orf142(Accession Q9BUH6), corresponding to amino acid residues P165-T204.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227622, 286257, 296565
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.