Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf156 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565709 Supplier Invitrogen™ Supplier No. PA565709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: STRSPHRPNAI GLTLAKLEKV EGGAIYLSGI DMIHGTPVLD IKPYIAEYDS PQNVMEPLAD FNLQNNQHTP NTVSQSDSKT DSCDQRQLSG CDEPQPHHST KRKPKCPED Highest antigen sequence identity to the following orthologs - mouse 65%, rat 70%.

S-adenosyl-L-methionine-dependent methyltransferase responsible for the addition of the methyl group in the formation of N6-methyl-N6-threonylcarbamoyladenosine at position 37 (m(6)t(6)A37) of the tRNA anticodon loop of tRNA(Ser)(GCU) (PubMed:25063302). The methyl group of m(6)t(6)A37 may improve the efficiency of the tRNA decoding ability. May bind to tRNA. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf156
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRMO
Gene Accession No. Q9BU70
Gene Alias 5830415F09Rik; AV014846; C9orf156; HSPC219; likely ortholog of H. sapiens chromosome 9 open reading frame 156 (C9orf156); NAP1; Nef (lentivirus myristoylated factor) associated protein 1; Nef associated protein 1; nef-associated protein 1; RGD1305420; RIKEN cDNA 5830415F09 gene; similar to Nef associated protein 1; thioesterase NAP1; TRMO; tRNA (adenine(37)-N6)-methyltransferase; tRNA methyltransferase O
Gene Symbols TRMO
Host Species Rabbit
Immunogen Recombinant Human C9orf156. Recombinant protein control fragment (Product #RP-106381).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.