Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf156 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985979
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5979 Supplier Bioss Supplier No. BS4210R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S-adenosyl-L-methionine-dependent methyltransferase responsible for the addition of the methyl group in the formation of N6-methyl-N6-threonylcarbamoyladenosine at position 37 (m(6)t(6)A37) of the tRNA anticodon loop of tRNA(Ser)(GCU). The methyl group of m(6)t(6)A37 may improve the efficiency of the tRNA decoding ability. May bind to tRNA.

Specifications

Antigen C9orf156
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene TRMO
Gene Accession No. Q9BU70
Gene Alias C9orf156; HSPC219; NAP1; Nef (lentivirus myristoylated factor) associated protein 1; Nef associated protein 1; nef-associated protein 1; RGD1305420; similar to Nef associated protein 1; thioesterase NAP1; Trmo; tRNA (adenine(37)-N6)-methyltransferase; tRNA methyltransferase O
Gene Symbols TRMO
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C9orf156.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51531
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.