Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C9orf16 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15316R

Description
C9orf16 is a protein coding gene.Specifications
C9orf16 | |
Polyclonal | |
Unconjugated | |
C9orf16 | |
C9orf16; chromosome 9 open reading frame 16; EST00098; UPF0184 protein C9orf16 | |
Rabbit | |
Protein A | |
RUO | |
79095 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9BUW7 | |
C9orf16 | |
KLH conjugated synthetic peptide derived from human C9orf16. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction