Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf171 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584478
Description
Immunogen sequence: AVKAGLVTAR ENLLYRQLND IRISDQDDRR MKKEPPPLPP NMTFGIRARP STPFFDLLQH RYLQLWVQEQ KATQKAIKL.
C9orf171 is a protein coding gene. Gene ontology (GO) annotation include cilium.
Specifications
C9orf171 | |
Polyclonal | |
Unconjugated | |
CFAP77 | |
1700101E01Rik; C9orf171; CFAP77; cilia and flagella associated protein 77; Cilia- and flagella-associated protein 77; FLJ46082; Gm823; RGD1564114; RIKEN cDNA 1700101E01 gene; similar to FLJ46082 protein; uncharacterized protein C9orf171; uncharacterized protein C9orf171 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZQR2 | |
CFAP77 | |
Recombinant protein corresponding to Human CFAP77. Recombinant protein control fragment (Product #RP-108220). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction