Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf173 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570192
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570192 Supplier Invitrogen™ Supplier No. PA570192
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%.

STPG3 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf173
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene STPG3
Gene Accession No. Q8N7X2
Gene Alias 4933433C11Rik; C9orf173; LOC499746; Protein STPG3; RIKEN cDNA 4933433C11 gene; similar to hypothetical gene supported by AK097565; BC033939; sperm-tail PG-rich repeat containing 3; Sperm-tail PG-rich repeat-containing protein 3; STPG3; uncharacterized protein C9orf173; uncharacterized protein C9orf173 homolog; uncharacterized protein LOC499746
Gene Symbols STPG3
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C9orf173.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441476
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.