Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf173 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983671
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3671 Supplier Bioss Supplier No. BS15321R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STPG3 is a protein coding gene.

Specifications

Antigen C9orf173
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STPG3
Gene Accession No. Q8N7X2
Gene Alias C9orf173; Protein STPG3; sperm-tail PG-rich repeat containing 3; Sperm-tail PG-rich repeat-containing protein 3; STPG3; uncharacterized protein C9orf173
Gene Symbols STPG3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C9orf173.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 441476
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.