Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562716
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562716 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562716 Supplier Invitrogen™ Supplier No. PA562716
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KYWLSQEEAD KCSPNYLGSD WYNTWRMEPY NSSCCNKYTT YLPRLPKEAR METAVRGMPL ECPPRPERLN AYEREVMVNM LN Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 67%.

This gene encodes a nuclear- or perinuclear-localized protein with no predicted domains or similarity to other known proteins. Expression of this gene is induced during the differentiation of bronchial epithelial cells, and the encoded protein may play a role in ciliogenesis. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf24
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C9orf24
Gene Accession No. Q8NCR6
Gene Alias bA573M23.4; C9orf24; CBE1; chromosome 9 open reading frame 24; ciliated bronchial epithelial protein 1; ciliated bronchial epithelium 1; NYD-SP22; SMRP1; Spermatid-specific manchette-related protein 1; testis development protein NYD-SP22
Gene Symbols C9orf24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf24. Recombinant protein control fragment (Product #RP-93029).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.