Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf40 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544688
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544688 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544688 Supplier Invitrogen™ Supplier No. PA544688
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: HNEEFWQYNT FQYWRNPLPP IDLADIEDLS EDTLTEATLQ GRNEGAEVDM Sequence homology: Horse: 79%; Human: 100%; Pig: 79%.

The function of C9orf40 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf40
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C9orf40
Gene Accession No. Q8IXQ3
Gene Alias C9orf40; chromosome 9 open reading frame 40; FLJ10110; uncharacterized protein C9orf40
Gene Symbols C9orf40
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C9orf40.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55071
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.