Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117820
This item is not returnable.
View return policy
Description
C9orf40 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
The function of C9orf40 remains unknown.
Specifications
C9orf40 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q8IXQ3 | |
C9orf40 | |
E. coli-derived Human C9orf40 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
C9orf40 | |
C9orf40; chromosome 9 open reading frame 40; FLJ10110; uncharacterized protein C9orf40 | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
55071 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction