Learn More
C9orf40 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304698AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function of C9orf40 remains unknown.Specifications
C9orf40 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8IXQ3, Q8VCE4 | |
C9orf40, D030056L22Rik | |
Between 75 and 125 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
C9orf40 | |
FLJ10110 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225995, 55071 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.