Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf41 Monoclonal Antibody (OTI6E4)
GREENER_CHOICE

Catalog No. PIMA525145
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA525145 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA525145 Supplier Invitrogen™ Supplier No. MA525145
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C9orf41 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf41
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone OTI6E4
Concentration 0.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CARNMT1
Gene Accession No. Q8N4J0
Gene Alias 2410127L17Rik; Anserine-producing methyltransferase; C1H9orf41; C9orf41; CARNMT1; Carnosine N-methyltransferase; carnosine N-methyltransferase 1; RGD1311863; RIKEN cDNA 2410127L17 gene; similar to RIKEN cDNA 2410127L17; UPF0586; UPF0586 protein C9orf41; UPF0586 protein C9orf41 homolog
Gene Symbols CARNMT1
Host Species Mouse
Immunogen Full length human recombinant protein of C9orf41 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138199
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.