Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf41 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555408
Description
Immunogen sequence: EEERLEREHF WKIINAFRYY GTSMHERVNR TERQFRSLPA NQQKLLPQFL LHLDKIRKCI DHNQEILLTI VNDCIHMFEN KEYGEDGNGK IMPASTFDMD KLKSTLKQFV RDWSETGKAE R Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 77%.
N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).
Specifications
| C9orf41 | |
| Polyclonal | |
| Unconjugated | |
| CARNMT1 | |
| 2410127L17Rik; Anserine-producing methyltransferase; C1H9orf41; C9orf41; CARNMT1; Carnosine N-methyltransferase; carnosine N-methyltransferase 1; RGD1311863; RIKEN cDNA 2410127L17 gene; similar to RIKEN cDNA 2410127L17; UPF0586; UPF0586 protein C9orf41; UPF0586 protein C9orf41 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 138199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N4J0 | |
| CARNMT1 | |
| Recombinant protein corresponding to Human C9orf41. Recombinant protein control fragment (Product #RP-89724). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction