Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf41 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983677
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3677 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3677 Supplier Bioss Supplier No. BS15329R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

N-methyltransferase that mediates the formation of anserine (beta-alanyl-N(Pi)-methyl-L-histidine) from carnosine. Anserine, a methylated derivative of carnosine (beta-alanyl-L-histidine), is an abundant constituent of vertebrate skeletal muscles. Also methylates other L-histidine-containing di- and tripeptides such as Gly-Gly-His, Gly-His and homocarnosine (GABA-His).

Specifications

Antigen C9orf41
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CARNMT1
Gene Accession No. Q8N4J0
Gene Alias Anserine-producing methyltransferase; C1H9orf41; C9orf41; Carnmt1; Carnosine N-methyltransferase; carnosine N-methyltransferase 1; RGD1311863; similar to RIKEN cDNA 2410127L17; UPF0586; UPF0586 protein C9orf41; UPF0586 protein C9orf41 homolog
Gene Symbols CARNMT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C9orf41.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138199
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.