Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf57 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570977
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570977 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570977 Supplier Invitrogen™ Supplier No. PA570977
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Human: 100%; Mouse: 83%; Rabbit: 83%.

C9orf57 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf57
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C9orf57
Gene Accession No. Q5W0N0
Gene Alias C9orf57; chromosome 9 open reading frame 57; uncharacterized protein C9orf57
Gene Symbols C9orf57
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C9orf57.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138240
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.