Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf61 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501734999 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4999 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4999 Supplier Proteintech Group Inc Supplier No. 256301AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C9orf61, also named as FAM189A2 and X123, is predomitly expressed in muscle. It has three isoforms with MW 45-50 kDa, 32-35 kDa and 27 kDa.

Specifications

Antigen C9orf61
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene FAM189A2
Gene Accession No. Q15884, Q4FZH1
Gene Alias C9orf61, FAM189A2, Protein FAM189A2, Protein X123, RP11 548B3.1, X123
Gene Symbols Fam189a2
Host Species Rabbit
Immunogen C9orf61 Fusion Protein Ag22242
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381217, 9413
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.