Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C9orf61 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15334R

Description
C9orf61, also named as FAM189A2 and X123, is predominantly expressed in muscle. It has three isoforms with MW 45-50 kDa, 32-35 kDa and 27 kDa.Specifications
C9orf61 | |
Polyclonal | |
Unconjugated | |
FAM189A2 | |
AI414343; AI666772; C9orf61; Fam189a2; family with sequence similarity 189 member A2; family with sequence similarity 189, member A2; Friedreich ataxia region gene X123; Gm967; Protein FAM189A2; Protein X123; X123 | |
Rabbit | |
Protein A | |
RUO | |
9413 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q15884 | |
FAM189A2 | |
KLH conjugated synthetic peptide derived from human C9orf61. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction