Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf64 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545087
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545087 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545087 Supplier Invitrogen™ Supplier No. PA545087
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: EMLSYGDRQE VEIRGCSLWC VELIRDCLLE LIEQKGEKPN GEINSILLDY Sequence homology: Cow: 79%; Dog: 90%; Human: 100%.

Involved in salvaging queuosine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf64
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C9orf64
Gene Accession No. Q5T6V5
Gene Alias C9orf64; chromosome 9 open reading frame 64; Queuosine salvage protein; UPF0553 protein; UPF0553 protein C9orf64
Gene Symbols C9orf64
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human C9orf64 (aa 266-315).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84267
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.