Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf66 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557352
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557352 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557352 Supplier Invitrogen™ Supplier No. PA557352
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RPTRLPRRLS PFWDPATCKN LEGGAGEVVR GRDPRRLRTS RSTEILGEDL AGPSAGAAAR PAAPPPQPRE PGA Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 35%.

C9orf66 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf66
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C9orf66
Gene Accession No. Q5T8R8
Gene Alias C9orf66; chromosome 9 open reading frame 66; uncharacterized protein C9orf66
Gene Symbols C9orf66
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf66. Recombinant protein control fragment (Product #RP-93074).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157983
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.