Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110797 Supplier Invitrogen™ Supplier No. PA5110797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NAIAFATGVL YGLSALGKKG DAISYARIQQ QRQQADEEKL AETLE Highest antigen sequence indentity to the following orthologs: Mouse - 98%, Rat - 98%.

Transmembrane protein which mediates synaptic endocytosis and fitness-based cell culling. In response to different stimulus strengths, controls two major modes of synaptic vesicle (SV) retrieval in hippocampal neurons; Clathrin- mediated endocytosis (CME) in response to mild stimulation and activity-dependent bulk endocytosis (ADBE) in response to strong stimulation (By similarity). In cytotoxic T-lymphoocytes (CTLs) facilitates calcium-dependent endocytosis of cytotoxic granules at the immuno synapse (By similarity). Different isoforms work as fitness fingerprints in 'loser' and 'winner' cells and thereby mediate win/lose decisions as part of the cell competition process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CACFD1
Gene Accession No. Q9UGQ2
Gene Alias 5930434B04Rik; C9orf7; CACFD1; calcium channel flower domain containing 1; calcium channel flower domain-containing protein 1; Calcium channel flower homolog; D9S2135; FLOWER; likely ortholog of H. sapiens chromosome 9 open reading frame 7 (C9orf7); mFwe; PSEC0107; PSEC0248; RGD1311501; UNQ3071/PRO9903
Gene Symbols CACFD1
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human C9orf7. Recombinant protein control fragment (Product #RP-91002).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11094
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.