Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ C9ORF72/C9RANT (poly-AP), Rat, Unlabeled, Clone: 14E2,

Catalog No. MABN1790MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1790MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1790MI Supplier MilliporeSigma™ Supplier No. MABN1790
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Expansion of a GGGGCC hexanucleotide repeat sequence in the non-coding region of human chromosome 9 open reading frame 72 or C9orf72 (also known as ALSFTD, FTDALS; Gene ID 203228) is the most common genetic abnormality in familial and sporadic frontotemporal dementia (FTD) and motor neuron disease (MND), with amyotrophic lateral sclerosis (ALS) as the most frequent form. The number of hexanucleotide repeats in the normal population ranges from 2 to 24, whereas up to several thousand repeats (700 - 4,400 repeats) are found in the pathologically expanded allele. Unconventional repeat-associated non-ATG-initiated translation (RANT) of the GGGGCC repeats in the three alternate reading frames from both sense and antisense transcripts generates five types of dipeptide repeat (DPR) protein species, each composed of repeating units of two amino acids, glycine-alanine (GA), glycine-proline (GP), glycine-arginine (GR), alanine-proline (AP), proline-arginine (PR), respectively. Immunofluorescence localization of exogenously epxressed DPR-GFP fusions in HEK293 transfectants showed dot-like or star-shaped poly-GA inclusions with predominant cytoplasmic and occasional nuclear localization, while poly-GR and poly-PR showed large dot-like intranuclear inclusions and occasional smaller cytoplasmic inclusions. In contrast, poly-PA was evenly distributed throughout the nucleus and cytoplasm without apparent aggregation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9ORF72/C9RANT (poly-AP)
Applications Western Blot
Classification Monoclonal
Clone 14E2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene C9orf72, ALSFTD, FTDALS
Gene Accession No. Q96LT7
Host Species Rat
Immunogen Recombinant GST fusion with a 10 AP-repeat sequence (GST-AP10).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone 14E2 recognizes C9ORF72/C9RANT (poly-AP) and other proteins containing poly-AP (poly-PA) sequence.
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.