Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C9ORF72/C9RANT (Poly-PR) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABN1354
Description
Anti-C9ORF72/C9RANT (Poly-PR) is a highly specific rabbit polyclonal antibody, that targets C9ORF72 (alpha-PR antisense) and has been tested in Dot Blot, Immunofluorescence, Immunohistochemistry, and Western Blotting.
Specifications
C9ORF72/C9RANT (Poly-PR) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Unpurified | |
RUO | |
Primary | |
This polyclonal antibody recognizes C9ORF72/C9RANT (poly-PR). | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. |
Immunofluorescence, Immunohistochemistry, Dot Blot, Western Blot | |
Please refer to lot specific datasheet. | |
C9orf72 | |
Ac-RPRPRPRPRPRPRPRPRC-amide. | |
100 μL | |
Neuroscience | |
NP_060795 | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction