Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C9orf72 Antibody (779), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313507
This item is not returnable.
View return policy
Description
C9orf72 Monoclonal antibody specifically detects C9orf72 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
C9orf72 | |
Monoclonal | |
Unconjugated | |
PBS, 20% Glycerol (pH7) | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
779 | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000 | |
ALSFTD, chromosome 9 open reading frame 72, FTDALS | |
The immunogen used to generate this antibody corresponds to human C9orf72. | |
100 μg | |
Neuroscience | |
203228 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction