Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf72 peptide 5 Rat anti-Human, Clone: 5F6, MilliporeSigma™

Catalog No. MABN2310100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN2310100 100 μg
MABN231025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABN2310100 Supplier MilliporeSigma Supplier No. MABN2310100UG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

C9orf72 peptide 5 Monoclonal antibody specifically detects C9orf72 peptide 5 in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf72 peptide 5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Purified rat monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Rat
Immunogen Ovalbumin-conjugated linear peptide corresponding to 20 amino acids from the N-terminal half (peptide 5) of human C9orf72.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.