Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C9orf72 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554905
Description
Immunogen sequence: YGLSIILPQT ELSFYLPLHR VCVDRLTHII RKGRIWMHKE RQENVQKIIL EGTERMEDQG QSIIPMLTGE VIPVMELLSS MKSHSVPEEI Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 94%.
C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.
Specifications
C9orf72 | |
Polyclonal | |
Unconjugated | |
C9orf72 | |
3110043O21Rik; AI840585; ALSFTD; C9orf72; chromosome 9 open reading frame 72; FTDALS; FTDALS1; Guanine nucleotide exchange C9orf72; Guanine nucleotide exchange C9orf72 homolog; protein C9orf72; protein C9orf72 homolog; RGD1359108; RIKEN cDNA 3110043O21 gene; similar to RIKEN cDNA 3110043O21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
203228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96LT7 | |
C9orf72 | |
Recombinant protein corresponding to Human C9orf72. Recombinant protein control fragment (Product #RP-92992). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction