Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106303
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106303 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106303 Supplier Invitrogen™ Supplier No. PA5106303
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total C9orf72.

C9orf72 may play a role in endosomal trafficking and autophagy. It is known to interact with HNRNPA1, HNRNPA2B1, UBQLN2, and small Rab GTPSases. C9orf72 localizes in the nucleus, cytoplasm, endosome, lysosome, and autophagosome. Mutations in this gene can result in frontotemporal dementia and/or amyotrophic lateral sclerosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf72
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene C9orf72
Gene Accession No. Q66HC3, Q96LT7
Gene Alias 3110043O21Rik; AI840585; ALSFTD; C9orf72; chromosome 9 open reading frame 72; FTDALS; FTDALS1; Guanine nucleotide exchange C9orf72; Guanine nucleotide exchange C9orf72 homolog; protein C9orf72; protein C9orf72 homolog; RGD1359108; RIKEN cDNA 3110043O21 gene; similar to RIKEN cDNA 3110043O21
Gene Symbols C9orf72, RGD1359108
Host Species Rabbit
Immunogen A synthesized peptide derived from human C9orf72(Accession Q96LT7), corresponding to amino acid residues P302-T352.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 203228, 313155, 73205
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.