Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C9orf72 (short form) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN1644MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN1644MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1644MI Supplier MilliporeSigma Supplier No. ABN1644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-C9ORF72 (short form) is a highly specific rabbit polyclonal antibody that targets short isoform of C9orf72 protein and has been tested in Immunofluorescence, Immunohistochemistry, Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf72 (short form)
Applications Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols C9orf72
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 12 amino acids from the C-terminal of short isoform (C9-S) of human C9orf72 with terminal asparagine substituted with lysine.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_659442
Test Specificity This polyclonal antibody specifically detects the short isoform (C9-S) of C9orf72 protein. It targets an epitope within 12 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.