Learn More
Invitrogen™ C9orf78 Polyclonal Antibody

Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, PC-12, Rat2. Predicted reactivity: Xenopus laevis (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | C9orf78 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | C9orf78 |
| Gene Accession No. | Q9NZ63 |
| Gene Alias | bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog |
| Gene Symbols | C9orf78, RGD1305178 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.