Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf78 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546104
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546104 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546104 Supplier Invitrogen™ Supplier No. PA546104
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: NRRDEDADMM KYIETELKKR KGIVEHEEQK VKPKNAEDCL YELPENIRVS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rat: 100%; Zebrafish: 79%.

Involved in the regulation of telomeric heterochromatin assembly and control of telomere length. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf78
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C9orf78
Gene Accession No. Q9NZ63
Gene Alias bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog
Gene Symbols C9orf78
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C9orf78.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51759
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.