Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C9orf78 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554361
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554361 Supplier Invitrogen™ Supplier No. PA554361
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPVVRKIFRR RRGDSESEED EQDSEEVRLK LEETREVQNL RKRPNGVSAV ALLVGEKVQE ETTLVDDPFQ MKTGGMVDMK KLKE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

Involved in the regulation of telomeric heterochromatin assembly and control of telomere length. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C9orf78
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C9orf78
Gene Accession No. Q9NZ63
Gene Alias bA409K20.3; C9orf78; chromosome 9 open reading frame 78; HCA59; Hepatocellular carcinoma-associated antigen 59; HSPC220; RGD1305178; similar to Hypothetical protein MGC11690; Telomere length and silencing protein 1 homolog; uncharacterized protein C9orf78; uncharacterized protein C9orf78 homolog
Gene Symbols C9orf78, RGD1305178
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C9orf78. Recombinant protein control fragment (Product #RP-92959).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 311855, 51759
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.